missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza A H1N1 (Hickox / 1940) Hemagglutinin Protein

A DNA sequence encoding the Influenza A virus (strain A/Hickox/1940 H1N1) Influenza A H1N1 (strain A/Hickox/1940) Hemagglutinin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7521-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16029160

Quantity Price
1 € 437.00 / 50µg
6 € 407.98 / 50µg
Estimated Shipment
04-11-2020
Add to basket


Specifications

Specifications

Influenza A H1N1 (Hickox / 1940) Hemagglutinin Protein
50 μg
Not determined.
Full Length
Influenza
His-SUMO
Tris-based buffer, 50% glycerol
Greater than 90% as determined by SDS-PAGE.
Research Use Only
HA
Recombinant Protein
E. coli
DTICIGYHAN NSTDTVDTVL EKNVTVTHSV NLLEDSHNGK LCRLKGIAPL QLGKCNIAGW ILGNPECESL LSKRSWSYIA ETPNSENGTC YPGDFADYEE LREQLSSVSS FERFEIFPKE RSWPNHNINI GVTAACSHAG KSSFYKNLLW LTEKDGSYPN LNKSYVNKKE KEVLVLWGVH HPSNIENQKT LYRKENAYVS VVSSNYNRRF TPEIAERPKV RGQAGRMNYY WTLLEPGDTI IFEANGNLIA PWYAFALSRG LGSGIITSNA SMDECDTKCQ TPQGAINSSL PFQNIHPFTI GECPKYVRST KLRMVTGLRN IPSIQS
Videos
SDS
Documents