missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza A H3N2 (Bangkok / 1 / 1979) Hemagglutinin Protein

A DNA sequence encoding the Influenza A virus (strain A/Bangkok/1/1979 H3N2) Influenza A H3N2 (strain A/Bangkok/1/1979) Hemagglutinin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9635-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16028551

  • € 3175.00 / 1mg


Estimated Shipment
29-10-2020
Add to basket

Specifications

Specifications

Influenza A H3N2 (Bangkok / 1 / 1979) Hemagglutinin Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Influenza
His
Tris-based buffer, 50% glycerol
Greater than 90% as determined by SDS-PAGE.
Research Use Only
HA
Recombinant Protein
Yeast
GIFGAIAGFI ENGWEGMXXG WYGFRHQNSE GTGQAADLKS TQAAIDQING KLNRVIEKTN EKFHQIEKEF SEVEGRIQDL EKYVEDTKID LWSYNAELLV ALENQHTIDL TDSEMNKLFE KTRRQLRENA EDMGNGCFKI YHKCDNACIG SIRNGTYDHD VYRDEALNNR FQIKGVELKX GYKDWILWIS FAISCFLLCV VLLGFIMVXC QKGNIRCNIC I
Videos
Documents