missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza-B Qingdao Protein
Click to view available options
Quantity:
1 mg
10 μg
50 μg
Unit Size:
10µg
1mg
50µg
Specifications
Specifications
| Name | Influenza-B Qingdao Protein |
| Quantity | 10 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Influenza |
| Species | Influenza B virus |
| Protein Tag | Untagged |
| Buffer | The B/Qingdao/102/91 solution contains 0.1M NaCl, 10mM Tris HCl, 1mM EDTA pH 8.0, 0.1% sodium azide (NaN3) and 0.005% thimerosal. |
| Purity or Quality Grade | Greater than 90.0% as determined byAnalysis by SDS-PAGE. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction