missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza H1N1 Taiwan Protein

Product Code. 15909308
Click to view available options
Quantity:
1 mg
10 μg
50 μg
Unit Size:
10µg
1mg
50µg
This item is not returnable. View return policy

Product Code. 15909308

Brand: enQuireBio™ QP1207950ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Name Influenza H1N1 Taiwan Protein
Quantity 50 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Influenza
Species Baculovirus
Protein Tag Untagged
Buffer The H1N1A/Taiwan/1/86 solution contains STE, 0.1% sodium azide (NaN3) and 0.005% thimerosal.
Purity or Quality Grade Greater than 90.0% as determined by SDS-PAGE.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.