missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza Matrix protein 1
A DNA sequence encoding the Influenza A virus (strain A/USA:Phila/1935 H1N1) Matrix protein 2, was expressed in the hosts and tags indicated.
€ 459.00 - € 1640.00
Specifications
Name | Influenza Matrix protein 1 |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | M2 |
Protein Length | Full Length Transmembrane Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16029101
|
enQuireBio™
QP9964-IV-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16039101
|
enQuireBio™
QP9964-IV-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16019101
|
enQuireBio™
QP9964-IV-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Influenza Matrix protein 1 | |
Not determined. | |
Full Length Transmembrane Protein | |
Influenza | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
M2 | |
Recombinant Protein | |
E. coli | |
MSLLTEVETP IRNEWGCRCN GSSDPLVIAA SIIGILHLIL WILDRLLFKC IYRRFKYGLK RGPSTEGVPE SMREEYRKEQ QSAVDADDGH FVNIEPE | |
Greater than 80% as determined by SDS-PAGE. |