missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant M. tuberculosis Immunogenic protein MPT64 Protein
A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) Immunogenic protein MPT64, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9654-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
WP_003409954.1 | |
50 μg | |
Not determined. | |
Full Length | |
M. tuberculosis | |
His | |
Tris-based buffer, 50% glycerol |
M. tuberculosis Immunogenic protein MPT64 Protein | |
Research Use Only | |
mpt64 | |
Recombinant Protein | |
Yeast | |
APKTYCEELK GTDTGQACQI QMSDPAYNIN ISLPSYYPDQ KSLENYIAQT RDKFLSAATS STPREAPYEL NITSATYQSA IPPRGTQAVV LKVYQNAGGT HPTTTYKAFD WDQAYRKPIT YDTLWQADTD PLPVVFPIVQ GELSKQTGQQ VSIAPNAGLD PVNYQNFAVT NDGVIFFFNP GELLPEAAGP TQVLVPRSAI DSMLA | |
Greater than 90% as determined by SDS-PAGE. |