missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant M. tuberculosis Low molecular weight antigen MTB12 Protein

A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) Low molecular weight antigen MTB12, was expressed in the hosts and tags indicated.

€ 352.00 - € 4075.00

Specifications

Accession Number WP_003917622.1
Name M. tuberculosis Low molecular weight antigen MTB12 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol mtb12
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16008581
View Documents Promotion Details
enQuireBio™
QP9684-YE-100UG
100 μg
€ 1100.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16028581
View Documents Promotion Details
enQuireBio™
QP9684-YE-1MG
1 mg
€ 4075.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16018581
View Documents Promotion Details
enQuireBio™
QP9684-YE-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16038581
View Documents Promotion Details
enQuireBio™
QP9684-YE-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16048581
View Documents Promotion Details
enQuireBio™
QP9684-YE-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16058581
View Documents Promotion Details
enQuireBio™
QP9684-YE-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

WP_003917622.1
Research Use Only
mtb12
Recombinant Protein
Yeast
DPASAPDVPT AAQLTSLLNS LADPNVSFAN KGSLVEGGIG GTEARIADHK LKKAAEHGDL PLSFSVTNIQ PAAAGSATAD VSVSGPKLSS PVTQNVTFVN QGGWMLSRAS AMELLQAAGN
Greater than 90% as determined by SDS-PAGE.
M. tuberculosis Low molecular weight antigen MTB12 Protein
Not determined.
Full Length
M. tuberculosis
His
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Product Certifications