missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant M. tuberculosis Low molecular weight antigen MTB12 Protein

A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) Low molecular weight antigen MTB12, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9684-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16028581

  • € 4075.00 / 1mg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

WP_003917622.1
1 mg
Not determined.
Full Length
M. tuberculosis
His
Tris-based buffer, 50% glycerol
M. tuberculosis Low molecular weight antigen MTB12 Protein
Research Use Only
mtb12
Recombinant Protein
Yeast
DPASAPDVPT AAQLTSLLNS LADPNVSFAN KGSLVEGGIG GTEARIADHK LKKAAEHGDL PLSFSVTNIQ PAAAGSATAD VSVSGPKLSS PVTQNVTFVN QGGWMLSRAS AMELLQAAGN
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers