Learn More
enQuireBio™ Recombinant Macaque CCL20 / MIP-3 alpha Protein
A DNA sequence encoding the Macaca mulatta (Rhesus macaque) CCL20 / MIP-3 alpha, was expressed in the hosts and tags indicated.
€ 309.00 - € 3405.00
Specifications
Accession Number | NP_001028026.1 |
---|---|
Gene ID (Entrez) | 574182 |
Name | CCL20 / MIP-3 alpha Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15917659
|
enQuireBio™
QP5772-EC-100UG |
100 μg |
€ 985.00
100µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15937659
|
enQuireBio™
QP5772-EC-1MG |
1 mg |
€ 3405.00
1mg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15927659
|
enQuireBio™
QP5772-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15947659
|
enQuireBio™
QP5772-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15957659
|
enQuireBio™
QP5772-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15967659
|
enQuireBio™
QP5772-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001028026.1 | |
CCL20 / MIP-3 alpha Protein | |
Not determined. | |
Full Length of Mature Protein | |
Macaque | |
His-GST | |
Tris-based buffer, 50% glycerol |
574182 | |
Research Use Only | |
CCL20 | |
Recombinant Protein | |
E. coli | |
ASNFDCCLRY TDRILHPKFI VGFTQQLANE TCDINAVVFH TKKGLSVCAN PKQTWVKLIV RRLSKKINKM | |
Greater than 90% as determined by SDS-PAGE. |