missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Macaque IL-1 alpha / IL1A / IL1F1 Protein

A DNA sequence encoding the Macaca mulatta (Rhesus macaque) IL-1 alpha / IL1A / IL1F1 Protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6213-ye-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16021630

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001036222.1
IL-1 alpha / IL1A / IL1F1 Protein
Research Use Only
IL1A
Recombinant Protein
Yeast
SAPFSFLSNM TYHFIRIIKH EFILNDTLNQ TIIRANDQHL TAAAIHNLDE AVKFDMGAYT SSKDDTKVPV ILRISKTQLY VSAQDEDQPV LLKEMPEINK TITGSETNFL FFWETHGTKN YFISVAHPNL FIATKHDNWV CLAKGLPSIT DFQILENQA
Greater than 90% as determined by SDS-PAGE.
700193
200 μg
Not determined.
Full Length
Macaque
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers