Learn More
R&D Systems™ Recombinant Monkeypox Virus A30L Fc Chimera Protein
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
Bioassay data are not available.
Specifications
Specifications
| Accession Number | NP_536567.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Molecular Weight (g/mol) | M.W. Observed: 52-60 kDa, under reducing conditions., M.W. Theoretical: 41 kDa |
| Quantity | 100 μg |
| Source | Human embryonic kidney cell, HEK293-derived monkeypox virus zaire-96-i-16 A30L protein MD Human IgG1 (Pro100-Lys330) IEGR MPXV A30L (Gln22-Leu146) (N-terminus) Accession # NP_536567.1 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Reconstitution | Reconstitute at 500 μg/mL in PBS. |
| Show More |