missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse ADH7 Protein
A DNA sequence encoding the Mus musculus (Mouse) ADH7, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7693-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_033756.2 | |
ADH7 Protein | |
Research Use Only | |
ADH7 | |
Recombinant Protein | |
E. coli | |
MGTAGKVIKC KAAVLWGVNQ PFSIEEIEVA PPKAKEVRVK ILATGICRTD DHIIKGSMVS KFPVIVGHEA VGVVESVGEG VTTVRPGDKV IPLFLPQCRE CNACLNPEGN LCIRSDLTGR GVLADGTTRF TCKGKPVQHF MNTSTFTEYT VLDESSVAKV DGAAPPEKAC LIGCGFSTGY GAAVKTAKVT PGSTCVVFGL GGVGLSVIMG CKAAGASRII GIDINKDKFQ KALAVGATEC ISPKDSTKPI SEVLSDMTGN TIQYTFEVIG RLETMVDALS SCHMNYGTSV VVGAPPSAKM LTYDPMLLFT GRTWKGCVFG GWKSRDDVPK LVTEFLEKKF DLDQLITHTL PFNNINEGFE LLYSGKSIRT VLTF | |
Greater than 90% as determined by SDS-PAGE. |
11529 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |