missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Alanine aminotransferase 1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Alanine aminotransferase 1, was expressed in the hosts and tags indicated.
€ 287.00 - € 4075.00
Specifications
Accession Number | NP_877957.1 |
---|---|
Gene ID (Entrez) | 76282 |
Name | Alanine aminotransferase 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16028881
|
enQuireBio™
QP9872-YE-100UG |
100 μg |
€ 920.00
100µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16048881
|
enQuireBio™
QP9872-YE-1MG |
1 mg |
€ 4075.00
1mg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16038881
|
enQuireBio™
QP9872-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058881
|
enQuireBio™
QP9872-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16068881
|
enQuireBio™
QP9872-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078881
|
enQuireBio™
QP9872-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_877957.1 | |
Alanine aminotransferase 1 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
76282 | |
Research Use Only | |
GPT | |
Recombinant Protein | |
Yeast | |
ASQRNDRIQA SRNGLKGKVL TLDTMNPCVR RVEYAVRGPI VQRALELEQE LRQGVKKPFT EVIRANIGDA QAMGQRPITF FRQVLALCVY PNLLSSPDFP EDAKRRAERI LQACGGHSLG AYSISSGIQP IREDVAQYIE RRDGGIPADP NNIFLSTGAS DAIVTMLKLL VAGEGRARTG VLIPIPQYPL YSAALAELDA VQVDYYLDEE RAWALDIAEL RRALCQARDR CCPRVLCVIN PGNPTGQVQT RECIEAVIRF AFEEGLFLMA DEVYQDNVYA EGSQFHSFKK VLTEMGPPYA TQQELASFHS VSKGYMGECG FRGGYVEVVN MDAEVQKQMA KLMSVRLCPP VPGQALMGMV VSPPTPSEPS FKQFQAERQE VLAELAAKAK LTEQVFNEAP GIRCNPVQGA MYSFPQIQLP LKAVQRAQDL GLAPDMFFCL CLLEETGICV VPGSGFGQQE GTYHFRMTIL PPMEKLRVLL EKLRHFHAKF THEYS | |
Greater than 90% as determined by SDS-PAGE. |