missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse alpha-Synuclein (Control) Protein, Pre-formed Fibrils

Product Code. 18218792 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
2 x 100 μg Vials
5 x 100 μg Vials
Unit Size:
1 set
200µg
500µg
This item is not returnable. View return policy

Product Code. 18218792

Brand: Novus Biologicals™ NBP261596100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified and high bioactivity. Generating reliable and reproducible results.

Active Mouse Recombinant Alpha Synuclein Protein Aggregate (pre-formed fibrils, Type 1). NCBI Accession #: NP_001035916.1. The Recombinant Mouse alpha-Synuclein (Control) Protein is derived from E. coli. The Recombinant Mouse alpha-Synuclein (Control) Protein has been validated for the following applications: Western Blot, In vitro assay, In vivo assay, SDS-Page, Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2 mg/ml
For Use With (Application) Bioactivity, In vitro Assay, In vivo Assay, SDS-PAGE, Western Blot
Formulation PBS
Gene ID (Entrez) 6622
Molecular Weight (g/mol) 14.46 kDa
Quantity 100 μg
Source E. coli
Storage Requirements Store at -80°C in the dark. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Symbol SNCA
Cross Reactivity alpha-Synuclein
Species Mouse
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.