missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Angiopoietin-like protein 1

A DNA sequence encoding the Mus musculus (Mouse) Angiopoietin-like protein 8, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7923-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031621

  • € 2195.00 / 500µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001074409.1
Angiopoietin-like protein 1
Research Use Only
Gm6484
Recombinant Protein
Yeast
VRPAPVAPLG GPEPAQYEEL TLLFHGALQL GQALNGVYRA TEARLTEAGH SLGLYDRALE FLGTEVRQGQ DATQELRTSL SEIQVEEDAL HLRAEATARS LGEVARAQQA LRDTVRRLQV QLRGAWLGQA HQEFETLKAR ADKQSHLLWA LTGHVQRQQR EMAEQQQWLR QIQQRLHTAA LPA
Greater than 90% as determined by SDS-PAGE.
624219
500 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers