missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse ANTXR1 Protein
A DNA sequence encoding the Mus musculus (Mouse) ANTXR1, was expressed in the hosts and tags indicated.
€ 137.00 - € 2450.00
Specifications
Accession Number | NP_473382.1 |
---|---|
Gene ID (Entrez) | 69538 |
Name | ANTXR1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15936719
|
enQuireBio™
QP5649-EC-10UG |
10 μg |
€ 137.00
10µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976719
|
enQuireBio™
QP5649-EC-50UG |
50 μg |
€ 339.00
50µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15926719
|
enQuireBio™
QP5649-EC-100UG |
100 μg |
€ 545.00
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956719
|
enQuireBio™
QP5649-EC-200UG |
200 μg |
€ 870.00
200µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966719
|
enQuireBio™
QP5649-EC-500UG |
500 μg |
€ 1500.00
500µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15946719
|
enQuireBio™
QP5649-EC-1MG |
1 mg |
€ 2450.00
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_473382.1 | |
ANTXR1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
69538 | |
Research Use Only | |
Antxr1 | |
Recombinant Protein | |
E. coli | |
EDGGPACYGG FDLYFILDKS GSVLHHWNEI YYFVEQLAHR FISPQLRMSF IVFSTRGTTL MKLTEDREQI RQGLEELQKV LPGGDTYMHE GFERASEQIY YENSQGYRTA SVIIALTDGE LHEDLFFYSE REANRSRDLG AIVYCVGVKD FNETQLARIA DSKDHVFPVN DGFQALQGII HSILKKSCIE ILAAEPSTIC AGESFQVVVR GNGFRHARNV DRVLCSFKIN DSVTLNEKPF AVEDTYLLCP APILKEVGMK AALQVSMNDG LSFISSSVII TTTHCSDGS | |
Greater than 90% as determined by SDS-PAGE. |