missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse ANXA5 Protein
A cDNA sequence encoding the ANXA5 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11035-20ug
Additional Details : Weight : 0.01000kg
Specifications
ANXA5 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSP MGSMATRGTV TDFPGFDGRA DAEVLRKAMK GLGTDEDSIL NLLTSRSNAQ RQEIAQEFKT LFGRDLVDDL KSELTGKFEK LIVAMMKPSR LYDAYELKHA LKGAGTDEKV LTEIIASRTP EELSAIKQVY EEEYGSNLED DVVGDTSGYY QRMLVVLLQA NRDPDTAIDD AQVELDAQAL FQAGELKWGT DEEKFITIFG TRSVSHLRRV FDKYMTISGF QIEETIDRET SGNLEQLLLA VVKSIRSIPA YLAETLYYAM KGAGTDDHTL IRVVVSRSEI DLFNIRKEFR KNFATSLYSM IKGDTSGDYK KALLLLCGGE DD | |
Greater than 95.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
ANXA5 protein solution (1 mg/ml) containing Phosphate Buffered Saline, 10% glycerol and 1mM DTT. |