missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Arid5a Protein

A DNA sequence encoding the Mus musculus (Mouse) Arid5a, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9789-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16028731

  • € 920.00 / 100µg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001165676.1
Arid5a Protein
Research Use Only
Arid5a
Recombinant Protein
Yeast
MAAPPAKGNT EQSEEGDLPQ LPVSPKPDDE QSRSQSPTQL QDSPEAGGEQ EEEQAFLVSL YKFMKERHTP IERVPHLGFK QINLWKIYKA VEKLGAYELV TGRRLWKNVY DELGGSPGST SAATCTRRHY ERLVLPYVRH LKGEDDKPLP PTKPRKQYKM AKELRGDDGT TEKLKKAKDS EERRVEQTTP GKTKSDATGQ TQLPCQGSSR DSTEQLGPVS GPSPPLTGAS SCPEAYKRLL SSFYCKGAHG IMSPLAKKKL LAQVSKAEAL QCQEEGCRHG ARSPNKDIQD SPQNLRGPAE NSEHQLTPRE GLQAPGGSTR MEAQVGPCPT APMFSGCFHA YPTEVLKPVS QHPRDFFSGL KDRVLLGPPG KEEGPTTKES HLVWGGDANH PSAFHKGSTR KRSFYPKPKA CWVSPMAKVP TERPGAPSPH PSSPGLGSKR GLEEEGFAHG GKKLRAVSPF LKEVDSKETG GKPAAPGLAV SCLLGPTPGP TPPEAYRGTM LRCPLNFTGS ADPLKGQASL PFSPLVIPAF PAHLLATTGS SPMAASLMHF PPTPYDAVLR NRLGPASSAW HMPPVTTYAA PHFFHLNTKL
Greater than 90% as determined by SDS-PAGE.
214855
100 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers