missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Bax Protein
A cDNA sequence encoding the Bax was constructed and used to recombinantly synthesize the protein.
€ 137.00 - € 7270.00
Specifications
Name | Bax Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15956248
|
enQuireBio™
QP11131-2UG |
2 μg |
€ 137.00
2µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15936248
|
enQuireBio™
QP11131-10UG |
10 μg |
€ 212.00
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15946248
|
enQuireBio™
QP11131-1MG |
1 mg |
€ 7270.00
1mg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Bax Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
Untagged | |
The protein contains 10mM Tris-HCL pH 8.0, 1mM EDTA and 250mM NaCl. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MDGSGEQLGS GGPTSSEQIM KTGAFLLQGF IQDRAGRMAG ETPELTLEQP PQDASTKKLS ECLRRIGDEL DSNMELQRMI ADVDTDSPRE VFFRVAADMF ADGNFNWGRV VALFYFASKL VLKALCTKVP ELIRTIMGWT LDFLRERLLV WIQDQGGWEG LLSYFGTPTW QTVTIFVAGV LTASLTIWKK MG | |
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |