missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse beta-NGF / Beta-NGF Protein
Recombinant Protein
Brand: enQuireBio™ QP5412-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_038637.1 | |
beta-NGF / Beta-NGF Protein | |
Research Use Only | |
NGF | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
18049 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MSSTHPVFHM GEFSVCDSVS VWVGDKTTAT DIKGKEVTVL AEVNINNSVF RQYFFETKCR ASNPVESGCR GIDSKHWNSY CTTTHTFVKA LTTDEKQAAW RFIRIDTACV CVLSRKATRR G | |
Greater than 95% as determined by SDS-PAGE. |