missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse C5a Protein

Product Code. 15984188
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
This item is not returnable. View return policy

Product Code. 15984188

Brand: enQuireBio™ QP105411mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the C5a was constructed and used to recombinantly synthesize the protein.

Specifications

Name C5a Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity The ED50 of Recombinant Mouse C5a as determined by its ability to induce N-acetyl-b-D-glucosaminidase release from differentiated U937 human histiocytic lymphoma cells was 5-20ng/ml.
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag Untagged
Sequence MGLWGILCLL IFLDKTWGQE QTYVISAPKI LRVGSSENVV IQVHGYTEAF DATLSLKSYP DKKVTFSSGY VNLSPENKFQ NAALLTLQPN QVPREESPVS HVYLEVVSKH FSKSKKIPIT YNNGILFIHT DKPVYTPDQS VKIRVYSLGD DLKPAKRETV LTFIDPEGSE VDIVEENDYT GIISFPDFKI PSNPKYGVWT IKANYKKDFT TTGTAYFEIK EYVLPRFSVS IELERTFIGY KNFKNFEITV KARYFYNKVV PDAEVYAFFG LREDIKDEEK QMMHKATQAA KLVDGVAQIS FDSETAVKEL SYNSLEDLNN KYLYIAVTVT ESSGGFSEEA EIPGVKYVLS PYTLNLVATP LFVKPGIPFS IKAQVKDSLE QAVGGVPVTL MAQTVDVNQE TSDLETKRSI THDTDGVAVF VLNLPSNVTV LKFEIRTDDP ELPEENQASK EYEAVAYSSL SQSYIYIAWT ENYKPMLVGE YLNIMVTPKS PYIDKITHYN YLILSKGKIV QYGTREKLFS STYQNINIPV TQNMVPSARL LVYYIVTGEQ TAELVADAVW INIEEKCGNQ LQVHLSPDEY VYSPGQTVSL DMVTEADSWV ALSAVDRAVY KVQGNAKRAM QRVFQALDEK SDLGCGAGGG HDNADVFHLA GLTFLTNANA DDSHYRDDSC KEILRSKRNL HLLRQKIEEQ AAKYKHSVPK KCCYDGARVN FYETCEERVA RVTIGPLCIR AFNECCTIAN KIRKESPHKP VQLGRIHIKT LLPVMKADIR SYFPESWLWE IHRVPKRKQL QVTLPDSLTT WEIQGIGISD NGICVADTLK AKVFKEVFLE MNIPYSVVRG EQIQLKGTVY NYMTSGTKFC VKMSAVEGIC TSGSSAASLH TSRPSRCVFQ RIEGSSSHLV TFTLLPLEIG LHSINFSLET SFGKDILVKT LRVVPEGVKR ESYAGVILDP KGIRGIVNRR KEFPYRIPLD LVPKTKVERI LSVKGLLVGE FLSTVLSKEG INILTHLPKG SAEAELMSIA PVFYVFHYLE AGNHWNIFYP DTLSKRQSLE KKIKQGVVSV MSYRNADYSY SMWKGASAST WLTAFALRVL GQVAKYVKQD ENSICNSLLW LVEKCQLENG SFKENSQYLP IKLQGTLPAE AQEKTLYLTA FSVIGIRKAV DICPTMKIHT ALDKADSFLL ENTLPSKSTF TLAIVAYALS LGDRTHPRFR LIVSALRKEA FVKGDPPIYR YWRDTLKRPD SSVPSSGTAG MVETTAYALL ASLKLKDMNY ANPIIKWLSE EQRYGGGFYS TQDTINAIEG LTEYSLLLKQ IHLDMDINVA YKHEGDFHKY KVTEKHFLGR PVEVSLNDDL VVSTGYSSGL ATVYVKTVVH KISVSEEFCS FYLKIDTQDI EASSHFRLSD SGFKRIIACA SYKPSKEEST SGSSHAVMDI SLPTGIGANE EDLRALVEGV DQLLTDYQIK DGHVILQLNS IPSRDFLCVR FRIFELFQVG FLNPATFTVY EYHRPDKQCT MIYSISDTRL QKVCEGAACT CVEADCAQLQ AEVDLAISAD SRKEKACKPE TAYAYKVRIT SATEENVFVK YTATLLVTYK TGEAADENSE VTFIKKMSCT NANLVKGKQY LIMGKEVLQI KHNFSFKYIY PLDSSTWIEY WPTDTTCPSC QAFVENLNNF AEDLFLNSCE
Buffer Mouse C5a was lyophilized from a concentrated (1 mg/ml) solution in 20mM PBS, pH 7.5 and 350mM NaCl.
Purity or Quality Grade Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.