missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CD207 / Langerin Protein

A DNA sequence encoding the Mus musculus (Mouse) CD207 / Langerin, was expressed in the hosts and tags indicated.

€ 245.00 - € 3405.00

Specifications

Accession Number NP_659192.2
Gene ID (Entrez) 246278
Name CD207 / Langerin Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16091401
View Documents Promotion Details
enQuireBio™
QP7887-EC-100UG
100 μg
€ 770.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16011411
View Documents Promotion Details
enQuireBio™
QP7887-EC-1MG
1 mg
€ 3405.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16001411
View Documents Promotion Details
enQuireBio™
QP7887-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16021411
View Documents Promotion Details
enQuireBio™
QP7887-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16031411
View Documents Promotion Details
enQuireBio™
QP7887-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16041411
View Documents Promotion Details
enQuireBio™
QP7887-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_659192.2
CD207 / Langerin Protein
Not determined.
Extracellular Domain
Mouse
His
Tris-based buffer, 50% glycerol
246278
Research Use Only
Cd207
Recombinant Protein
E. coli
QAVFYPRLMG KILDVKSDAQ MLKGRVDNIS TLGSDLKTER GRVDDAEVQM QIVNTTLKRV RSQILSLETS MKIANDQLQI LTMSWGEVDS LSAKIPELKR DLDKASALNT KVQGLQNSLE NVNKLLKQQS DILEMVARGW KYFSGNFYYF SRTPKTWYSA EQFCISRKAH LTSVSSESEQ KFLYKAADGI PHWIGLTKAG SEGDWYWVDQ TSFNKEQSRR FWIPGEPNNA GNNEHCANIR VSALKCWNDG PCDNTFLFIC KRPYVQTTE
Greater than 80% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications