missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse CXCL5/ENA-78 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
1mg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
Description
CXCL5 Protein is fully biologically active when compared to standard. The biological activity determined by a chemoattract bioassay using human neutrophils is in a concentration range of 10-100 ng/mL.
Specifications
Specifications
| Accession Number | P50228, P50228, P50228 |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 6374 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 7.6 kDa |
| Quantity | 1mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/μg of CXCL5/ENA-78 as determined by LAL method. |
| Gene Alias | chemokine (C-C motif) ligand 5, C-X-C motif chemokine 5, ENA-78, ENA-78(1-78), Epithelial-derived neutrophil-activating protein 78, LIX/CXCL5, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78 |
| Gene Symbol | CXCL5 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction