missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Decorin Protein
Click to view available options
Quantity:
100 μg
missing translation for 'unitSize'
100µg
Description
The Recombinant Mouse Decorin Protein is derived from NS0. The Recombinant Mouse Decorin Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 13179 |
| Molecular Weight (g/mol) | 39 kDa |
| Name | Decorin |
| Quantity | 100 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse Decorin protein Gly17-Lys354 & Asp31-Lys354,with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction