missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse DLL4 Protein
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
Specifications
Specifications
| Name | DLL4 Protein |
| Quantity | 2 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Mouse |
| Species | Baculovirus |
| Protein Tag | His |
| Sequence | GSGIFQLRLQ EFVNQRGMLA NGQSCEPGCR TFFRICLKHF QATFSEGPCT FGNVSTPVLG TNSFVVRDKN SGSGRNPLQL PFNFTWPGTF SLNIQAWHTP GDDLRPETSP GNSLISQIII QGSLAVGKIW RTDEQNDTLT RLSYSYRVIC SDNYYGESCS RLCKKRDDHF GHYECQPDGS LSCLPGWTGK YCDQPICLSG CHEQNGYCSK PDECICRPGW QGRLCNECIP HNGCRHGTCS IPWQCACDEG WGGLFCDQDL NYCTHHSPCK NGSTCSNSGP KGYTCTCLPG YTGEHCELGL SKCASNPCRN GGSCKDQENS YHCLCPPGYY GQHCEHSTLT CADSPCFNGG SCRERNQGSS YACECPPNFT GSNCEKKVDR CTSNPCANGG QCQNRGPSRT CRCRPGFTGT HCELHISDCA RSPCAHGGTC HDLENGPVCT CPAGFSGRRC EVRITHDACA SGPCFNGATC YTGLSPNNFV CNCPYGFVGS RCEFPVGLPP SFPWVAHHHH HH |
| Buffer | DLL4 protein solution (1 mg/ml) contains Phosphate buffered saline (pH 7.4) and 10% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction