Learn More
enQuireBio™ Recombinant Mouse FLT3L / Flt3 ligand Protein
Recombinant Protein
Brand:  enQuireBio™ QP5405-2ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15936059
  • € 94.45 / 2µg
Estimated Shipment: 30-01-2023
to see stock.
Add to basket
Specifications
NP_038548.3
FLT3L / Flt3 ligand Protein
2 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Shows bioactivity at a concentration of 10 ng/mL in the OCI-AML5 cell proliferation assay. The bioactivity acceptance criteria is set at 1 x 10^5 units/mg.
Mouse
Untagged
10 mM sodium phosphate, pH 7.5
14256
Greater than 95% as determined by SDS-PAGE.
Research Use Only
Flt3l
Recombinant Protein
E. coli
MTPDCYFSHS PISSNFKVKF RELTDHLLKD YPVTVAVNLQ DEKHCKALWS LFLAQRWIEQ LKTVAGSKMQ TLLEDVNTEI HFVTSCTFQP LPECLRFVQT NISHLLKDTC TQLLALKPCI GKACQNFSRC LEVQCQPDSS TLLPPRSPIA LEATELPEPR PRQ
Videos
SDS
Documents
Certificates
Special Offers
Special Offers