missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Fragile histidine triad / FHIT Protein

A DNA sequence encoding the Mus musculus (Mouse) Fragile histidine triad / FHIT, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7503-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16019040

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001295215.1
Fragile histidine triad / FHIT Protein
Research Use Only
Fhit
Recombinant Protein
E. coli
SFRFGQHLIK PSVVFLKTEL SFALVNRKPV VPGHVLVCPL RPVERFRDLH PDEVADLFQV TQRVGTVVEK HFQGTSITFS MQDGPEAGQT VKHVHVHVLP RKAGDFPRND NIYDELQKHD REEEDSPAFW RSEKEMAAEA EALRVYFQA
Greater than 90% as determined by SDS-PAGE.
14198
10 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers