missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Fragile histidine triad / FHIT Protein

A DNA sequence encoding the Mus musculus (Mouse) Fragile histidine triad / FHIT, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7503-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16029040

  • € 2645.00 / 1mg


Estimated Shipment
02-11-2020
Add to basket

Specifications

Specifications

NP_001295215.1
Fragile histidine triad / FHIT Protein
1 mg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
14198
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Fhit
Recombinant Protein
E. coli
SFRFGQHLIK PSVVFLKTEL SFALVNRKPV VPGHVLVCPL RPVERFRDLH PDEVADLFQV TQRVGTVVEK HFQGTSITFS MQDGPEAGQT VKHVHVHVLP RKAGDFPRND NIYDELQKHD REEEDSPAFW RSEKEMAAEA EALRVYFQA
Videos
SDS
Documents