missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Galactocerebrosidase Protein

A DNA sequence encoding the Mus musculus (Mouse) Galactocerebrosidase, was expressed in the hosts and tags indicated.

€ 535.00 - € 1830.00

Specifications

Accession Number NP_032105.2
Gene ID (Entrez) 14420
Name Galactocerebrosidase Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16029131
View Documents Promotion Details
enQuireBio™
QP9977-IV-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16039131
View Documents Promotion Details
enQuireBio™
QP9977-IV-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16019131
View Documents Promotion Details
enQuireBio™
QP9977-IV-100UG
100 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_032105.2
Galactocerebrosidase Protein
Not determined.
Full Length
Mouse
Untagged
Tris-based buffer, 50% glycerol
14420
Research Use Only
GALC
Recombinant Protein
E. coli
YVLDDSDGLG REFDGIGAVS GGGATSRLLV NYPEPYRSEI LDYLFKPNFG ASLHILKVEI GGDGQTTDGT EPSHMHYELD ENYFRGYEWW LMKEAKKRNP DIILMGLPWS FPGWLGKGFS WPYVNLQLTA YYVVRWILGA KHYHDLDIDY IGIWNERPFD ANYIKELRKM LDYQGLQRVR IIASDNLWEP ISSSLLLDQE LWKVVDVIGA HYPGTYTVWN AKMSGKKLWS SEDFSTINSN VGAGCWSRIL NQNYINGNMT STIAWNLVAS YYEELPYGRS GLMTAQEPWS GHYVVASPIW VSAHTTQFTQ PGWYYLKTVG HLEKGGSYVA LTDGLGNLTI IIETMSHQHS MCIRPYLPYY NVSHQLATFT LKGSLREIQE LQVWYTKLGT PQQRLHFKQL DTLWLLDGSG SFTLELEEDE IFTLTTLTTG RKGSYPPPPS SKPFPTNYKD DFNVEYPLFS EAPNFADQTG VFEYYMNNED REHRFTLRQV LNQRPITWAA DASSTISVIG DHHWTNMTVQ CDVYIETPRS GGVFIAGRVN KGGILIRSAT GVFFWIFANG SYRVTADLGG WITYASGHAD VTAKRWYTLT LGIKGYFAFG MLNGTILWKN VRVKYPGHGW AAIGTHTFEF AQFDNFRVEA AR
Greater than 85% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications