Learn More
enQuireBio™ Recombinant Mouse Glypican-4 Protein
Product Code. p-7152790
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16040870 10 μg 10µg
16080870 50 μg 50µg
16030870 100 μg 100µg
16060870 200 μg 200µg
16070870 500 μg 500µg
16050870 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 16040870 Supplier enQuireBio™ Supplier No. QP6110ye10ug
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
A DNA sequence encoding the Mus musculus (Mouse) Glypican-4, was expressed in the hosts and tags indicated.
Specifications
Accession Number NP_032176.2
Gene ID (Entrez) 14735
Name Glypican-4 Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Gpc4
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Mouse
Species Yeast
Protein Tag GST
Sequence AGGAHAEAKP YLLAALCILF LAVQGEWR
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.