missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Guanine nucleotide-binding protein G(o) subunit alpha Protein
A DNA sequence encoding the Mus musculus (Mouse) Guanine nucleotide-binding protein G(o) subunit alpha, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6102-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001106855.1 | |
Guanine nucleotide-binding protein G(o) subunit alpha Protein | |
Research Use Only | |
Gnao1 | |
Recombinant Protein | |
E. coli | |
GCTLSAEERA ALERSKAIEK NLKEDGISAA KDVKLLLLGA GESGKSTIVK QMKIIHEDGF SGEDVKQYKP VVYSNTIQSL AAIVRAMDTL GVEYGDKERK TDSKMVCDVV SRMEDTEPFS AELLSAMMRL WGDSGIQECF NRSREYQLND SAKYYLDSLD RIGAGDYQPT EQDILRTRVK TTGIVETHFT FKNLHFRLFD VGGQRSERKK WIHCFEDVTA IIFCVALSGY DQVLHEDETT NRMHESLMLF DSICNNKFFI DTSIILFLNK KDLFGEKIKK SPLTICFPEY PGSNTYEDAA AYIQTQFESK NRSPNKEIYC HMTCATDTNN IQVVFDAVTD IIIANNLRGC GLY | |
Greater than 90% as determined by SDS-PAGE. |
14681 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |