missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Guanine nucleotide-binding protein G(o) subunit alpha Protein

A DNA sequence encoding the Mus musculus (Mouse) Guanine nucleotide-binding protein G(o) subunit alpha, was expressed in the hosts and tags indicated.

€ 245.00 - € 2645.00

Specifications

Accession Number NP_001106855.1
Gene ID (Entrez) 14681
Name Guanine nucleotide-binding protein G(o) subunit alpha Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16090810
View Documents Promotion Details
enQuireBio™
QP6102-YE-100UG
100 μg
€ 770.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16030820
View Documents Promotion Details
enQuireBio™
QP6102-YE-500UG
500 μg
€ 2195.00
500µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16000820
View Documents Promotion Details
enQuireBio™
QP6102-YE-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16020820
View Documents Promotion Details
enQuireBio™
QP6102-YE-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16040820
View Documents Promotion Details
enQuireBio™
QP6102-YE-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16010820
View Documents Promotion Details
enQuireBio™
QP6102-YE-1MG
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_001106855.1
Guanine nucleotide-binding protein G(o) subunit alpha Protein
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
14681
Research Use Only
Gnao1
Recombinant Protein
Yeast
GCTLSAEERA ALERSKAIEK NLKEDGISAA KDVKLLLLGA GESGKSTIVK QMKIIHEDGF SGEDVKQYKP VVYSNTIQSL AAIVRAMDTL GVEYGDKERK TDSKMVCDVV SRMEDTEPFS AELLSAMMRL WGDSGIQECF NRSREYQLND SAKYYLDSLD RIGAGDYQPT EQDILRTRVK TTGIVETHFT FKNLHFRLFD VGGQRSERKK WIHCFEDVTA IIFCVALSGY DQVLHEDETT NRMHESLMLF DSICNNKFFI DTSIILFLNK KDLFGEKIKK SPLTICFPEY PGSNTYEDAA AYIQTQFESK NRSPNKEIYC HMTCATDTNN IQVVFDAVTD IIIANNLRGC GLY
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications