missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse H2-D1 Protein

A DNA sequence encoding the Mus musculus (Mouse) H2-D1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7403-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16028700

  • € 1705.00 / 500µg


Estimated Shipment
22-10-2020
Add to basket

Specifications

Specifications

H2-D1 Protein
500 μg
Not determined.
Extracellular Domain
Mouse
His
Tris-based buffer, 50% glycerol
Greater than 90% as determined by SDS-PAGE.
Research Use Only
H2-D1
Recombinant Protein
Yeast
GSHSLRYFVT AVSRPGFGEP RYMEVGYVDN TEFVRFDSDA ENPRYEPRAR WIEQEGPEYW ERETRRAKGN EQSFRVDLRT ALRYYNQSAG GSHTLQWMAG CDVESDGRLL RGYWQFAYDG CDYIALNEDL KTWTAADMAA QITRRKWEQA GAAERDRAYL EGECVEWLRR YLKNGNATLL RTDPPKAHVT HHRRPEGDVT LRCWALGFYP ADITLTWQLN GEELTQEMEL VETRPAGDGT FQKWASVVVP LGKEQKYTCH VEHEGLPEPL TLRWGKEEPP SSTKTNT
Videos
Documents