missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Hadh, mitochondrial Protein
A DNA sequence encoding the Mus musculus (Mouse) Hadh, mitochondrial, was expressed in the hosts and tags indicated.
€ 245.00 - € 3405.00
Specifications
Accession Number | NP_032238.2 |
---|---|
Gene ID (Entrez) | 15107 |
Name | Hadh, mitochondrial Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16000181
|
enQuireBio™
QP7685-EC-100UG |
100 μg |
€ 770.00
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16020181
|
enQuireBio™
QP7685-EC-1MG |
1 mg |
€ 3405.00
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16010181
|
enQuireBio™
QP7685-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16030181
|
enQuireBio™
QP7685-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16040181
|
enQuireBio™
QP7685-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16050181
|
enQuireBio™
QP7685-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_032238.2 | |
Hadh, mitochondrial Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
15107 | |
Research Use Only | |
Hadh | |
Recombinant Protein | |
E. coli | |
SSSSSASAAA KKILIKHVTV IGGGLMGAGI AQVAAATGHT VVLVDQTEDI LAKSKKGIEE SLKRMAKKKF TENPKAGDEF VEKTLSCLST STDAASVVHS TDLVVEAIVE NLKLKNELFQ RLDKFAAEHT IFASNTSSLQ ITNIANATTR QDRFAGLHFF NPVPMMKLVE VIKTPMTSQK TFESLVDFCK TLGKHPVSCK DTPGFIVNRL LVPYLIEAVR LHERGDASKE DIDTAMKLGA GYPMGPFELL DYVGLDTTKF ILDGWHEMEP ENPLFQPSPS MNNLVAQKKL GKKTGEGFYK YK | |
Greater than 90% as determined by SDS-PAGE. |