missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Hmgcs1, cytoplasmic Protein

A DNA sequence encoding the Mus musculus (Mouse) Hmgcs1, cytoplasmic, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7817-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16030981

  • € 600.00 / 100µg


Estimated Shipment
23-10-2020
Add to basket

Specifications

Specifications

NP_001278368.1
Hmgcs1, cytoplasmic Protein
100 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
208715
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Hmgcs1
Recombinant Protein
Yeast
MPGSLPLNAE ACWPKDVGIV ALEIYFPSQY VDQAELEKYD GVDAGKYTIG LGQARMGFCT DREDINSLCL TVVQKLMERH SLSYDCIGRL EVGTETIIDK SKSVKSNLMQ LFEESGNTDI EGIDTTNACY GGTAAVFNAV NWVESSSWDG RYALVVAGDI AIYATGNARP TGGVGAVALL IGPNAPLIFD RGLRGTHMQH AYDFYKPDML SEYPVVDGKL SIQCYLSALD RCYSVYRKKI RAQWQKEGKD KDFTLNDFGF MIFHSPYCKL VQKSLARMFL NDFLNDQNRD KNSIYSGLEA FGDVKLEDTY FDRDVEKAFM KASSELFNQK TKASLLVSNQ NGNMYTSSVY GSLASVLAQY SPQQLAGKRV GVFSYGSGLA ATLYSLKVTQ DATPGSALDK ITASLCDLKS RLDSRTCVAP DVFAENMKLR EDTHHLANYI PQCSIDSLFE GTWYLVRVDE KHRRTYARRP FTNDHSLDEG MGLVHSNTAT EHIPSPAKKV PRLPATSAES ESAVISNGEH
Videos
Documents