missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse HSP70 / HSPA1A Protein
A DNA sequence encoding the Mus musculus (Mouse) HSP70 / HSPA1A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6185-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_034608.2 | |
HSP70 / HSPA1A Protein | |
Research Use Only | |
Hspa1b | |
Recombinant Protein | |
E. coli | |
AKNTAIGIDL GTTYSCVGVF QHGKVEIIAN DQGNRTTPSY VAFTDTERLI GDAAKNQVAL NPQNTVFDAK RLIGRKFGDA VVQSDMKHWP FQVVNDGDKP KVQVNYKGES RSFFPEEISS MVLTKMKEIA EAYLGHPVTN AVITVPAYFN DSQRQATKDA GVIAGLNVLR IINEPTAAAI AYGLDRTGKG ERNVLIFDLG GGTFDVSILT IDDGIFEVKA TAGDTHLGGE DFDNRLVSHF VEEFKRKHKK DISQNKRAVR RLRTACERAK RTLSSSTQAS LEIDSLFEGI DFYTSITRAR FEELCSDLFR GTLEPVEKAL RDAKMDKAQI HDLVLVGGST RIPKVQKLLQ DFFNGRDLNK SINPDEAVAY GAAVQAAILM GDKSENVQDL LLLDVAPLSL GLETAGGVMT ALIKRNSTIP TKQTQTFTTY SDNQPGVLIQ VYEGERAMTR DNNLLGRFEL SGIPPAPRGV PQIEVTFDID ANGILNVTAT DKSTGKANKI TITNDKGRLS KEEIERMVQE AERYKAEDEV QRDRVAAKNA LESYAFNMKS AVEDEGLKGK LSEADKKKVL DKCQEVISWL DSNTLADKEE FVHKREELER VCSPIISGLY QGAGAPGAGG FGAQAPPKGA SGSGPTIEEV D | |
Greater than 90% as determined by SDS-PAGE. |
15511 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |