missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse IFN-gamma Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
Description
Interferon gamma Protein is fully biologically active when compared to standard. The ED50 as determined by an anti-viral assay using murine L929 cells infected with encephalomyocarditis (EMC) virus is less than 0.8 ng/mL, corresponding to a specific activity of >1.3 x 106IU/mg.
Specifications
Specifications
| Accession Number | P01580, P01580, P01580 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered solution in 20 mM Tris-HCl pH 8.0, 300 mM NaCl, 0.05 % Tween-20. |
| Gene ID (Entrez) | 3458 |
| Molecular Weight (g/mol) | 15.5 kDa |
| Quantity | 1 mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of IFN-gamma as determined by LAL method. |
| Gene Alias | IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction