missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse IFN-gamma Protein

Product Code. 18487401 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
This item is not returnable. View return policy

Product Code. 18487401

missing translation for 'mfr': Novus Biologicals™ NBP235071100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Interferon gamma Protein is fully biologically active when compared to standard. The ED50 as determined by an anti-viral assay using murine L929 cells infected with encephalomyocarditis (EMC) virus is less than 0.8 ng/mL, corresponding to a specific activity of >1.3 x 106IU/mg.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01580, P01580, P01580
Concentration LYOPH
For Use With (Application) PAGE, Bioactivity
Formulation Lyophilized from a 0.2 um filtered solution in 20 mM Tris-HCl pH 8.0, 300 mM NaCl, 0.05 % Tween-20.
Gene ID (Entrez) 3458
Molecular Weight (g/mol) 15.5 kDa
Quantity 100 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/ug of IFN-gamma as determined by LAL method.
Gene Alias IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
Gene Symbol IFNG
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled 20 mM Tris-HCl pH8.0 with 50 mM NaCl to a concentration of 0.1-0.3 mg/ml. Apportion stock solutions into working aliquots and store at <-20°C.
Research Category Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers
Purity or Quality Grade >95% by SDS-PAGE and HPLC
Protein IFN-gamma
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.