missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse IFN-kappa Protein

Product Code. 18798807
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in Sodium Acetate, NaCl and EDTA with BSA as a carrier protein
Lyophilized from a 0.2μm filtered solution in Sodium Acetate, NaCl and EDTA.
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18798807

Brand: R&D Systems™ 8437MK010/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse IFN-kappa Protein is derived from NS0. The Recombinant Mouse IFN-kappa Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q7TSL0
Formulation Lyophilized from a 0.2μm filtered solution in Sodium Acetate, NaCl and EDTA.
Gene ID (Entrez) 56832
Molecular Weight (g/mol) 21kDa
Name IFN-kappa
Preparation Method Reconstitute at 200μg/mL in sterile water.
Quality Control Testing SDS-PAGE: 19-22kDa, reducing conditions
Quantity 10 μg
Source Mouse myeloma cell line,NS0-derived mouse IFN-kappa protein Leu22-Lys199
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Leu22
Without Additives Carrier Protein
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE with silver staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.