missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse IgG Protein
Shop All Bio Techne Products
Click to view available options
:
50μg; Unlabeled
Description
An un-tagged recombinant protein corresponding to the amino acids 98-330 of Mouse IgG The Recombinant Mouse IgG Protein is derived from Baculovirus. The Recombinant Mouse IgG Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| For Use With (Application) | SDS-PAGE |
| Formulation | Liquid. In 20mM Tris-HCl buffer (pH 8.0) |
| Molecular Weight (g/mol) | 26.6kDa |
| Name | Mouse IgG Protein |
| Purification Method | SDS-PAGE |
| Storage Requirements | 4°C short term, −20°C long term. Avoid freeze-thaw cycles. |
| Cross Reactivity | Mouse |
| Purity or Quality Grade | >90% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction