missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse IgG Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
0.5 mg
100μg
Unit Size:
0.50mg
100µg
Description
A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 98-330 of mouse Mouse IgG The Recombinant Mouse IgG Protein is derived from Baculovirus. The Recombinant Mouse IgG Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Concentration | 1mg/mL |
| Formulation | 20mM Tris-HCl buffer (pH8.0) containing 10% glycerol |
| Quantity | 100μg |
| Storage Requirements | Store at 4° short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO - research use only |
| Recombinant | Recombinant |
| Purity or Quality Grade | >90%, by SDS-PAGE |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction