missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IL-17AF Protein

Product Code. 15975669
Click to view available options
Quantity:
1 mg
100 μg
25 μg
5 μg
500 μg
Unit Size:
100µg
1mg
25µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 15975669

Brand: enQuireBio™ QP52055ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Name IL-17AF Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol IL17
Biological Activity Mouse IL-17AF bioactivity was tested by measuring IL-6 production in 3T3 cells. IL-17AF activity was measurable at concentrations above 50 ng/ml.
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag Untagged
Sequence IL-17A:MAA IIPQSSACPN TEAKDFLQNV KVNLKVFNSL GAKVSSRRPS DYLNRSTSPW TLHRNEDPDR YPSVIWEAQC RHQRCVNAEG KLDHHMNSVL IQQEILVLKR EPESCPFTFR VEKMLVGVGC TCVASIVRQA A IL-17F:MR KNPKAGVPAL QKAGNCPPLE DNTVRVDIRI FNQNQGISVP REFQNRSSSP WDYNITRDPH RFPSEIAEAQ CRHSGCINAQ GQEDSTMNSV AIQQEILVLR REPQGCSNSF RLEKMLLKVG CTCVKPIVHQ AA
Buffer Sterile filtered (0.22 um) solution containing 0.1% TFA
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.