missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Il27 alpha Protein
Recombinant Protein
€ 112.00 - € 7420.00
Specifications
Accession Number | NP_663611.1 |
---|---|
Gene ID (Entrez) | 246779 |
Name | Il27 alpha Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15966169
|
enQuireBio™
QP5457-2UG |
2 μg |
€ 112.00
2µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15946169
|
enQuireBio™
QP5457-10UG |
10 μg |
€ 263.00
10µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15936169
|
enQuireBio™
QP5457-100UG |
100 μg |
€ 985.00
100µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976169
|
enQuireBio™
QP5457-500UG |
500 μg |
€ 4515.00
500µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956169
|
enQuireBio™
QP5457-1MG |
1 mg |
€ 7420.00
1mg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_663611.1 | |
Il27 alpha Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MALVALSQPR VQCHASRYPV AVDCSWTPLQ APNSTRSTSF IATYRLGVAT QQQSQPCLQR SPQASRCTIP DVHLFSTVPY MLNVTAVHPG GASSSLLAFV AERIIKPDPP EGVRLRTAGQ RLQVLWHPPA SWPFPDIFSL KYRLRYRRRG ASHFRQVGPI EATTFTLRNS KPHAKYCIQV SAQDLTDYGK PSDWSLPGQV ESAPHKP | |
Greater than 95% as determined by SDS-PAGE. |
246779 | |
Research Use Only | |
IL27Ra | |
Mouse | |
Untagged | |
10 mM sodium bicarbonate, pH 8.5 |