missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse KLK-8 / Kallikrein-8 Protein

A DNA sequence encoding the Mus musculus (Mouse) KLK-8 / Kallikrein-8, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9804-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16028761

  • € 770.00 / 100µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001311327.1
KLK-8 / Kallikrein-8 Protein
Research Use Only
Klk8
Recombinant Protein
Yeast
ILEGRECIPH SQPWQAALFQ GERLICGGVL VGDRWVLTAA HCKKQKYSVR LGDHSLQSRD QPEQEIQVAQ SIQHPCYNNS NPEDHSHDIM LIRLQNSANL GDKVKPVQLA NLCPKVGQKC IISGWGTVTS PQENFPNTLN CAEVKIYSQN KCERAYPGKI TEGMVCAGSS NGADTCQGDS GGPLVCDGML QGITSWGSDP CGKPEKPGVY TKICRYTTWI KKTMDNRD
Greater than 94.7% as determined by SDS-PAGE.
259277
100 μg
Not determined.
Full. Length. of. Mature. Protein.
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers