missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse LDHB Protein

Product Code. 15975038
Click to view available options
Quantity:
1 μg
5 μg
50 μg
Unit Size:
1µg
50µg
5µg
This item is not returnable. View return policy

Product Code. 15975038

Brand: enQuireBio™ QP107451ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the LDHB was constructed and used to recombinantly synthesize the protein.

Specifications

Name LDHB Protein
Quantity 1 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity Specific activity is > 150 units/mg, in which one unit will 1.0 umole of pyruvate to L-lactate and beta-NAD per minute at pH 7.5.
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MGSMATLKEK LIASVADDEA AVPNNKITVV GVGQVGMACA ISILGKSLAD ELALVDVLED KLKGEMMDLQ HGSLFLQTPK IVADKDYSVT ANSKIVVVTA GVRQQEGESR LNLVQRNVNV FKFIIPQIVK YSPDCTIIVV SNPVDILTYV TWKLSGLPKH RVIGSGCNLD SARFRYLMAE KLGIHPSSCH GWILGEHGDS SVAVWSGVNV AGVSLQELNP EMGTDNDSEN WKEVHKMVVD SAYEVIKLKG YTNWAIGLSV ADLIESMLKN LSRIHPVSTM VKGMYGIENE VFLSLPCILN ARGLTSVINQ KLKDDEVAQL RKSADTLWDI QKDLKDL
Buffer LDHB protein solution (1 mg/ml) containing 20mM Tris-HCl (pH 8.0), 1mM DTT and 10% glycerol.
Purity or Quality Grade Greater than 95.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.