missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Leptin Protein
Recombinant Protein
Brand: enQuireBio™ QP5429-5mg
Additional Details : Weight : 0.01000kg
Specifications
NP_032519.1 | |
Leptin Protein | |
Research Use Only | |
LEP | |
Mouse | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |
16846 | |
5 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MVPIQKVQDD TKTLIKTIVT RINDISHTQS VSAKQRVTGL DFIPGLHPIL SLSKMDQTLA VYQQVLTSLP SQNVLQIAND LENLRDLLHL LAFSKSCSLP QTSGLQKPES LDGVLEASLY STEVVALSRL QGSLQDILQQ LDVSPEC | |
Greater than 95% as determined by SDS-PAGE. |