Learn More
enQuireBio™ Recombinant Mouse Leptin Protein
Recombinant Protein
Brand:  enQuireBio™ QP5429-1000ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15936109
  • € 230.00 / 1mg
Estimated Shipment: 30-01-2023
to see stock.
Add to basket
Specifications
NP_032519.1
Leptin Protein
1 mg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Recombinant Protein
E. coli
MVPIQKVQDD TKTLIKTIVT RINDISHTQS VSAKQRVTGL DFIPGLHPIL SLSKMDQTLA VYQQVLTSLP SQNVLQIAND LENLRDLLHL LAFSKSCSLP QTSGLQKPES LDGVLEASLY STEVVALSRL QGSLQDILQQ LDVSPEC
16846
Greater than 95% as determined by SDS-PAGE.
Research Use Only
LEP
Mouse
Untagged
10 mM sodium phosphate, pH 7.5
Videos
SDS
Documents
Certificates
Special Offers
Special Offers