missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse LIF Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
1mg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
Description
LIF protein is fully biologically active when compared to standard. The specific activity is determined by inducing differentiation of murine M1 myeloid leukemic cells. The minimum detectable concentration of LIF in this assay is 0.01 ng/mL. The specific activity of >1.0 x 10^8 IU/mg, where 50 units is defined as the amount of rMuLIF required to induce differentiation in 50 % of the M1 colonies in 1 mL agar cultures
Specifications
Specifications
| Accession Number | P09056, P09056, P09056 |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB, pH 7.4, with 0.02 % TWEEN 20. |
| Gene ID (Entrez) | 3976 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 19.9 kDa |
| Quantity | 5 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/μg of LIF as determined by LAL method. |
| Gene Alias | CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI |
| Gene Symbol | LIF |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction