missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse LSECtin/CLEC4G Protein
Description
Measured by its binding ability in a functional ELISA. When Recombinant Mouse BTNL4 Fc Chimera (Catalog # 9590-BT ) is immobilized at 2 μg/mL (100 μL/well), Recombinant Mouse LSECtin/CLEC4G (Catalog # 10363-CL) binds with an ED50 of 0.15-1.2 μg/mL.
Specifications
Specifications
| Accession Number | Q8BNX1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 339390 |
| Molecular Weight (g/mol) | M.W. (Observed): 36-44 kDa, under reducing conditions; M.W. (theoretical): 29 kDa |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived mouse LSECtin/CLEC4G protein Hemagglutinin Tag (YPYDVPDYA) Mouse LSECtin/CLEC4G (Leu52-Tyr294) (N-terminus) Accession # Q8BNX1 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?