missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Midkine Protein
A cDNA sequence encoding the Midkine was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10786-20ug
Additional Details : Weight : 0.01000kg
Specifications
Midkine Protein | |
Research Use Only | |
Fully biologically active when compared to standard. Determined by its ability to chemoattract human neutrophils using a concentration range of 10-100 ng/ml corresponding to a specific activity of 10,000-100,000IU/mg. | |
Mouse | |
Untagged | |
Lyophilized from a 0.2um filtered concentrated solution in PBS, pH 7.4. |
20 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
VAKKKEKVKK GSECSEWTWG PCTPSSKDCG MGFREGTCGA QTQRVHCKVP CNWKKEFGAD CKYKFESWGA CDGSTGTKAR QGTLKKARYN AQCQETIRVT KPCTSKTKSK TKAKKGKGKD | |
Greater than 95.0% as determined by HPLC and SDS-PAGE. |